missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2/Her2 Antibody (rERBB2/9401), Novus Biologicals™
Mouse Monoclonal Antibody
2150.00 NOK - 4160.00 NOK
Specifications
Antigen | ErbB2/Her2 |
---|---|
Clone | rERBB2/9401 |
Concentration | 0.2 mg/mL |
Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18664064
|
Novus Biologicals
NBP3-23594-100ug |
100 μg |
4160.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18692654
|
Novus Biologicals
NBP3-23594-20ug |
20 μg |
2150.00 NOK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ErbB2/Her2 Monoclonal antibody specifically detects ErbB2/Her2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ErbB2/Her2 | |
0.2 mg/mL | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Phospho Specific, Protein Kinase, Stem Cell Markers, Tumor Suppressors | |
10 mM PBS with 0.05% BSA | |
2064 | |
IgG2b κ | |
Protein A purified |
rERBB2/9401 | |
Immunohistochemistry-Paraffin : 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
Recombinant fragment (around aa1155-1255) of human ErbB2/Her2 protein (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ErbB2/Her2 Antibody (rERBB2/9401), Novus Biologicals™