Learn More
Eppendorf Reference™ 2, Mechanical Single-Channel Pipette
Shop All Eppendorf Products
Click to view available options
Volume (Metric):
0.1 to 2.5 μL
0.25 to 2.5 mL
0.5 to 10 μL
0.5 to 5 mL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Color:
Blue
Dark Gray
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
Increments:
0.002 μL
0.01 μL
0.02 μL
0.1 μL
0.2 μL
1 μL
2 μL
5 μL
10 μL
Unit Size:
Each
Description
- Integrated pipetting system with internal RFID technology
- Serial number is printed on multiple components of pipette
- Color coded and volume labeled pipette for quick identification of volume size/tip size
- Round upper part of pipette makes it possible to work in every position
- Four-digit display for exact, quick and secure volume setting (volume lock included)
- Spring-loaded tip cone reduces effort during operation/ejection
- Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust
- High flexibility with ability to remove single cones
- Ensures continuous identical pipet alignment during work process
- Fully autoclavable pipet with smooth surface and few rough edges
- Single-button operation
- Has Spring-loaded tip cone, movable parts, stainless steel external edges with customizable labeling area
- Ergonomic handling with reduced operating effort
- Simple and quick tip ejection with active aerosol reduction
- Reduce repetitive strain injuries by not having to move your thumb sideways
- Haptic feedback before tip ejection
- High precision and accuracy provides reliable pipetting results
- User friendly secondary adjustment
Greener Choice Claims
- Environmental benefits include:
- Free-of a substance that causes environmental harm
- Non-Toxic
- Ozone-Safe or Safe for the Ozone Layer
- Renewable Energy
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Orange |
| For Use With | Pipette Tip 300 μL |
| Number of Channels | 1 |
| Product Line | Reference 2 |
| Type | Pipette |
| Volume (Metric) | 30 to 300 μL |
| For Use With (Equipment) | 300 μL pipette tips |
| Includes | epT.I.P.S.™ Box 2.0 with 96 Tips |
| Increments | 0.2 μL |
| Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 15
Videos
-
Eppendorf Reference 2 - Simple Adjustment
-
Eppendorf Reference 2 - Eppendorf TrackIT
-
Eppendorf Reference 2 - The New Legend
-
Eppendorf Reference 2 - A flight through the new legend
Product Content Correction
Product Title
Eppendorf Reference™ 2, Mechanical Single-Channel Pipette >