Learn More
Fisherbrand™ EPA Vial Kit, 24-400 screw thread, Clear Glass, White PP screw cap, Center hole, Silicone/PTFE septum
Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
20 mL
40 mL
60 mL
Material (Liner):
Natural Silicone/Beige PTFE
Natural Silicone/PTFE
White Silicone/Beige PTFE
Unit Size:
Pack of 100
Specifications
Specifications
| Material (Liner) | White Silicone/Beige PTFE |
| Color | Clear |
| Diameter (Metric) Outer | 27.5 mm |
| Material | Glass |
| Volume (Metric) | 60 mL |
| For Use With (Application) | TOC (EPA) |
| Thread Style | 24-400 |
| Closure Material | Polypropylene |
| Closure Type | Screw Cap |
| Height (Metric) | 140 mm |
| Show More |
Product Content Correction
Product Title
Fisherbrand™ EPA Vial Kit, 24-400 screw thread, Clear Glass, White PP screw cap, Center hole, Silicone/PTFE septum >