Learn More
Memmert™ Entry Port 38 mm for UN30
Optional entry port for Memmert Ovens.
Brand: Memmert™ UN30F7
This item is not returnable.
View return policy
Description
- Entry port with 38 mm clear diameter
- Flap and stopper closure
- Custom position TBC in the back
Specifications
UN30 Oven | |
Entry Port | |
38 mm |
Product Content Correction
Product Title
Memmert™ Entry Port 38 mm for UN30 > For Use With: UN30