Learn More
Memmert™ Entry Port 23 mm for UN30, Custom TBC Right Side
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UN30F5
Product Code. 17580086
  • 2235.00 NOK / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Description
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in right side
Specifications
Specifications
UN30 Oven
Entry Port
23 mm
Product Title
Memmert™ Entry Port 23 mm for UN30, Custom TBC Right Side > For Use With: UN30