Learn More
Memmert™ Entry Port 23 mm, Standard Position Side Right Side Centre/Top, For Hcp50
Use these entry ports with convection ovens. Memmert™ Convection and Universal Oven Entry Points can be closed by flap in special positions at the back.
Brand:  Memmert™ HCP50F3
Product Code. 16384877
  • 4895.00 NOK / Each
Only null left
Estimated Shipment: 19-08-2025
Add to basket
Description
Description
  • Entry port with clear diameter
  • For Memmert Ovens
Specifications
Specifications
Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
Stainless Steel
Entry Port
23 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Entry Port 23 mm, Standard Position Side Right Side Centre/Top, For Hcp50 >