-
Sartorius Entris II Advanced Line application peakhold
-
Sartorius Entris II Advanced Line application statistic
-
Sartorius Entris II Advanced Line application mixing
-
Sartorius Entris II Advanced Line application weighing
-
Sartorius Entris II Advanced Line application percentage weighing
-
Sartorius Entris II Advanced Line application conversion
-
Sartorius Entris II Advanced Line application components
-
Sartorius Entris II Advanced Line application counting
-
Sartorius Entris II Advanced Line application pipette smart test
-
Sartorius Entris II Advanced Line application animal weighing
Sartorius Entris™ II Advanced Precision Balances with Country-Specific Verification
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
Brand: Sartorius BCA12201I-1CEU
42949.50 NOK valid until 2025-12-31
Use promo code "24377" to get your promotional price.
This item is not returnable.
View return policy
Description
- Entris™ II Advanced Balances - Quality, Value, and Consistency
- For accurate weighing results that can be trusted time after time
- Simplifies balance leveling with built-in, real-time level support
- Increases reliability of weighing results using integrated protection systems
- Controlled access to balance settings with user management
- Gapless documentation thanks to CalAuditTrail
Specifications
CE Certificate | |
Verified | |
Intuitive Graphic Touch Technology | |
Glass, PBT, Stainless Steel | |
≤0.9 seconds | |
0.2 A |
D-sub connector, 9-pin; USB UTL; USB Device | |
12,200 g | |
100 mg | |
Electronic Precision Balance | |
100/240 V (± 10%) | |
50/60 Hz |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Sartorius Entris II Advanced Line application peakhold
-
Sartorius Entris II Advanced Line application statistic
-
Sartorius Entris II Advanced Line application mixing
-
Sartorius Entris II Advanced Line application weighing
-
Sartorius Entris II Advanced Line application percentage weighing
-
Sartorius Entris II Advanced Line application conversion
-
Sartorius Entris II Advanced Line application components
-
Sartorius Entris II Advanced Line application counting
-
Sartorius Entris II Advanced Line application pipette smart test
-
Sartorius Entris II Advanced Line application animal weighing
CE_16695633.pdf
File size: 1.2 MB
CE Certificate
File size: 1.2 MB
Accessories and Services
Optional Accessory
-
Sartorius™ Weighing Tables WEIGHING TABLE Product Code: 10529341
32230.00 NOK / Each -
Sartorius Printer Paper
X5 Paper rolls Sartorius for data printe... Product Code: 10663182304.00 NOK / Pack of 5 -
Sartorius Premium GLP Lab Printer for Secura™ and Quintix&t... Printer GLP, with RS232 & USB cables & p... Product Code: 14811441
13195.00 NOK / Each -
Sartorius Printer Paper, Labels, and Ink for YDP Printers X5 Standard thermal paper rolls of 24 m Product Code: 15318824
335.00 NOK / Pack of 5 -
Sartorius™ Kensington Lock Cable for Laboratory Balances Kensington-Lock anti-theft device Product Code: 17751828
840.00 NOK / Each -
Sartorius™ Y-connector for Laboratory Balances Y-Connector RS232 9-pin (M) > 2 × 9-pin ... Product Code: 17761818
1035.00 NOK / Each
Product Content Correction
Product Title
Sartorius Entris™ II Advanced Precision Balances with Country-Specific Verification > 12,200 g; 0.1 g; Verified