Learn More
Sartorius Entris™ II Advanced Precision Balances with Country-Specific Verification
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
Brand:  Sartorius BCA4202I-1CEU
33041.00 NOK valid until 2025-12-31
Use promo code "24377" to get your promotional price.
 View more versions of this product
Product Code. 16685663
  • 34780.00 NOK / Each
Description
  • Entris™ II Advanced Balances - Quality, Value, and Consistency
  • For accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in, real-time level support
  • Increases reliability of weighing results using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE Certificate
Verified
Intuitive Graphic Touch Technology
Glass, PBT, Stainless Steel
≤0.9 seconds
0.2 A
D-sub connector, 9-pin; USB UTL; USB Device
4200 g
10 mg
Electronic Precision Balance
100/240 V (± 10%)
50/60 Hz
Videos
  • Sartorius Entris II Advanced Line application statistic
    Video Player is loading.
    Current Time 0:00
    Duration 1:18
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:18
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    Sartorius Entris II Advanced Line application statistic
  • Sartorius Entris II Advanced Line application peakhold
    Video Player is loading.
    Current Time 0:00
    Duration 0:34
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:34
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Sartorius Entris II Advanced Line application peakhold
  • Sartorius Entris II Advanced Line application components
    Video Player is loading.
    Current Time 0:00
    Duration 1:19
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:19
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    Sartorius Entris II Advanced Line application components
  • Sartorius Entris II Advanced Line application weighing
    Video Player is loading.
    Current Time 0:00
    Duration 0:35
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:35
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Sartorius Entris II Advanced Line application weighing
  • Sartorius Entris II Advanced Line application mixing
    Video Player is loading.
    Current Time 0:00
    Duration 1:21
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:21
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    Sartorius Entris II Advanced Line application mixing
  • Sartorius Entris II Advanced Line application conversion
    Video Player is loading.
    Current Time 0:00
    Duration 0:54
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:54
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    Sartorius Entris II Advanced Line application conversion
  • Sartorius Entris II Advanced Line application percentage weighing
    Video Player is loading.
    Current Time 0:00
    Duration 1:06
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:06
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    Sartorius Entris II Advanced Line application percentage weighing
  • Sartorius Entris II Advanced Line application counting
    Video Player is loading.
    Current Time 0:00
    Duration 1:08
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:08
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    Sartorius Entris II Advanced Line application counting
  • Sartorius Entris II Advanced Line application density determination
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Sartorius Entris II Advanced Line application density determination
    • Sartorius Entris II Advanced Line application pipette smart test
      Video Player is loading.
      Current Time 0:00
      Duration 1:31
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 1:31
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • default, selected
      Sartorius Entris II Advanced Line application pipette smart test
    SDS
    Documents
    Documents
    Brochures
    Product Guides
    Certificates
    View accessory products
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Sartorius Entris™ II Advanced Precision Balances with Country-Specific Verification > 4,200 g; 10 mg; Verified