missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENPP6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
3505.00 NOK - 5500.00 NOK
Specifications
Antigen | ENPP6 |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18365563
|
Bio-Techne
NBP3-17051-25UL |
25 μg |
3505.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18329654
|
Bio-Techne
NBP3-17051-100UL |
100 μg |
5500.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ENPP6 Polyclonal antibody specifically detects ENPP6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ENPP6 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS, pH 7.2, 40% glycerol | |
133121 | |
IgG | |
Affinity purified |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
B830047L21Rik, EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 6, ectonucleotide pyrophosphatase/phosphodiesterase family member 6, E-NPP 6, MGC33971, NPP6, NPP-6 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: KYISLNDLQQVKDRGPVVSLWPAPGKHSEIYNKLSTVEHMTVYEKEAIPSRFYYKKGKFVSPLTLVADEGWFITENREMLPFWMNSTGRREGWQRG | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title