Learn More
Merck RC800 Mini-Reservoir
Accessory for simplified stirred cell operation
Brand:  Merck 6028
Product Code. 10783291
  • 29625.00 NOK / Each
Only null left
Estimated Shipment: 28-07-2025
Description
  • Graduated acrylic body
  • Acetal cover, removable for easy cleaning
  • Acetal fittings, Buna-N, silicone O-rings
  • Safety valve for slow pressure bleed (max. pressure: 5 bar)
  • Polyethylene tubing
  • Wrap-around clamp for extra safety
  • Chemically sanitizable
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Merck RC800 Mini-Reservoir > RC800 Mini-Reservoir; 19H x 12D x 13cm W