Learn More
Cole-Parmer™ Embosser Tape for Manual Laboratory Label Maker
Embosser Tape for the Manual Laboratory Label Maker sticks firmly to glass, plastic, metal, rubber, and wood.
Brand: Cole-Parmer™ 65543-48
This item is not returnable.
View return policy
Description
- Easily remove back adhesive shield
- Choose from four colors to color-code your lab
Specifications
Colored Label Tape | |
118 in. | |
300 cm | |
118 x 3/8 in. | |
Non-sterile | |
Label |
Blue | |
3/8 in. | |
0.95 cm | |
Peel and Stick | |
Monochrome |