Learn More
Motic™ ELWD Condenser for AE2000
Extra-long working distance condenser suitable for phase contrast. Motic™ ELWD Condenser for AE2000, when used with phase accessories, facilitates visualization of transparent specimens.
Brand:  Motic™ 1101000201621
Product Code. 11792348
  • 3640.00 NOK / Each
Only null left
Estimated Shipment: 28-08-2025
Description
  • Concentrates and focuses light passing through the specimen.
  • Extra-long working-distance condenser.
  • Suitable for phase contrast.
Specifications
Specifications
NA: 0.30
ELWD Condenser
AE2000 Microscopes
72 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Motic™ ELWD Condenser for AE2000 > WD: 72mm