Learn More
Bibby Scientific™ Electrothermal™ Replacement Heater
Brand:  Bibby Scientific™ 10086-49
 View more versions of this product
Product Code. 11778432
  • 6585.00 NOK / Each
Only null left
Estimated Shipment: 03-07-2025
Specifications
2000 mL
EMA
500w
Replacement Heater
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bibby Scientific™ Electrothermal™ Replacement Heater > Capacity: 2000mL; For Use With: EMA