missing translation for 'onlineSavingsMsg'
Learn More

EIF2AK2 Monoclonal Antibody (OTI3F5), TrueMAB™, OriGene

Mouse Monoclonal Antibody

4065.15 NOK

Specifications

Antigen EIF2AK2
Clone OTI3F5
Concentration 1 mg/mL
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17578114
Encompass_Preferred
View Documents Promotion Details
Origene Technologies
TA813289S
30 μL
4065.15 NOK
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17558114
Encompass_Preferred
View Documents Promotion Details
Origene Technologies
TA813289
100 μL
4065.15 NOK
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EIF2AK2 Monoclonal antibody specifically detects EIF2AK2 in Human samples. It is validated for Immunohistochemistry, Western Blot
Specifications

Specifications

EIF2AK2
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
2310047A08Rik; 4732414G15Rik; AI467567; AI747578; double stranded RNA activated protein kinase; dsRNA-activated kinase; EC 2.7.11.1; eIF-2 alpha; eIF2a Kinase; eIF-2A protein kinase 2; EIF2AK1; EIF2AK2; eukaryotic translation initiation factor 2 alpha kinase 2; eukaryotic translation initiation factor 2-alpha kinase 2; IFN- type I-induced and dsRNA-activated kinase; IFN-induced and double-stranded RNA-activated kinase; interferon-induced, double-stranded RNA-activated protein kinase; interferon-inducible elF2alpha kinase; interferon-inducible RNA-dependent protein kinase; MGC126524; OTTHUMP00000201320; P1/eIF-2A protein kinase; p68 kinase; PKR; PPP1R83; Prkr; protein kinase R; protein kinase RNA-activated; protein kinase, interferon inducible double stranded RNA dependent; Protein kinase, interferon-inducible double stranded RNA dependent; protein phosphatase 1, regulatory subunit 83; serine/threonine-protein kinase TIK; T-cell viral integration site; Tik; Tyrosine-protein kinase EIF2A
Human recombinant protein fragment corresponding to amino acids 51-191 of human EIF2AK2 (NP_001129123) produced in E.coli.
Primary
-20° C, Avoid Freeze/Thaw Cycles
EIF2AK2
OTI3F5
Immunohistochemistry, Western Blot
Unconjugated
Mouse
Human
P19525
5610
IgG1
Protein A/G
Antibody
Videos
SDS
Documents

Documents

Product Certifications