Learn More
Cole Parmer™ Economical Polypropylene Graduated Cylinder
Polypropylene (PP) cylinders offer very good chemical resistance.
Brand: Cole Parmer™ 06137-72
This item is not returnable.
View return policy
Description
- Meet ISO Standard 6706.
- Calibrated to contain for measuring final volumes.
- Molded graduations for durability.
- Polypropylene construction provides very good chemical resistance.
Specifications
Polypropylene | |
ISO Standard 6706 |
Product Content Correction
Product Title
Cole Parmer™ Economical Polypropylene Graduated Cylinder > 25 mL