Learn More
Jenway™ Economical Flame Photometer
Economical Flame Photometer features an automatic gas shut-off that ensures operator safety.
Brand:  Jenway™ 83055-02
Product Code. 12461250
  • 54770.00 NOK / Each
Only null left
Estimated Shipment: 23-07-2025
Description
  • Unit comes with Na+, K+, Li+, Ca+, and Ba+ filters
Specifications
Specifications
115 V
50/60 Hz
Economical Flame Photometer
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Jenway™ Economical Flame Photometer > Power: 115 VAC