Learn More
Velp Scientifica™ ECO Condenser Test Tube
Velp Scientifica™ ECO Condenser Test Tube with 3-meter polythylene tube for sludge analysis.
Brand:  Velp Scientifica™ CA0091667
1275.17 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12303317
  • 1315.00 NOK / Each
Focus On Clean Water
Use promo code 24139 at checkout
View Available Promotions
Only null left
Estimated Shipment: 14-08-2025
Description
  • For use with: ECO 8, ECO 16, ECO 25 Thermoreactor
Specifications
Specifications
200 mm
ECO 8, ECO 16, ECO 25 Thermoreactor
3-meter polythylene tube
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Shop now for up to 60% off Clean Water Research essentials
    Promo Code: 24139
    Expires2025-08-21
    New Offer
    Save Now
Product Content Correction
Product Title
Velp Scientifica™ ECO Condenser Test Tube > For Use With: ECO 8, ECO 16, ECO 25 Thermoreactor