Learn More
Sharpsguard™ Eco Orange Com-Plus Waste Container
The new Sharpsguard™ eco orange containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have not been contaminated with medicines.
Brand: Sharpsguard™ DD479OLR
This item is not returnable.
View return policy
Description
- Sharpsguard™ eco orange com-plus is rectangular for easy storage in medical bags, it has a sliding door with an aperture which will take a 20 mL syringe.
- This container is already supplied with the lid assembled. Details about how to lock the container and keep the users and contents safe can be found on the label.
- It is designed to fit safely and securely into our POUDS™ tray range and other carrying trays.
- This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-02.
Specifications
0.78 L | |
Yellow | |
1 Ea. |
Orange | |
204 x 68 x 120 mm | |
159 g |