Learn More
STEMCELL Technologies EasySep™ Non-Human Primate B Cell Isolation Kit
Immunomagnetic isolation of untouched non-human primate B cells
Brand:  STEMCELL Technologies 100-0345
Encompass
Product Code. 17127571
  • 16751.97 NOK / 1 set
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Non-Human Primate B Cell Isolation Kit is designed to isolate untouched and highly purified B cells from fresh or previously frozen non-human primate peripheral blood mononuclear cells (PBMCs) in as little as 20 minutes by immunomagnetic negative selection
  • The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
  • Isolated cells are immediately available for downstream applications such as flow cytometry, culture, or DNA/RNA extraction
  • This kit has been verified for use with rhesus and cynomolgus macaques
Specifications
Specifications
B Cells
Cell Isolation
1 x 109 cells