Learn More
STEMCELL Technologies EasySep™ Mouse Neutrophil Enrichment Kit
Immunomagnetic isolation of untouched mouse neutrophils
Brand:  STEMCELL Technologies 19762
Encompass
Product Code. 17177951
  • 10868.48 NOK / 1 set
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Mouse Neutrophil Enrichment Kit is designed to isolate neutrophils from single-cell suspensions of bone marrow or other tissues by negative selection
  • Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-neutrophils
  • Labeled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotin and dextran
  • These cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
Specifications
Specifications
Granulocytes and Subsets
Cell Isolation
1 x 109 Cells