Learn More
STEMCELL Technologies EasySep™ Human CD33 Positive Selection Kit II
Immunomagnetic positive selection of human CD33+ cells (e.g. myeloid cells)
Brand: STEMCELL Technologies 17876
This item is not returnable.
View return policy
Description
- The EasySep™ Human CD33 Positive Selection Kit II is designed to isolate CD33+ cells from ammonium chloride lysed blood or bone marrow, or from nucleated cells prepared by density separation with RosetteSep™ DM-M density medium (Catalog #15725) by positive selection
- Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD33 and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- The CD33 antigen is expressed on cells of the myeloid lineage
- This product replaces the EasySep™ Human CD33 Positive Selection Kit (Catalog #18257) for even faster cell isolations
Specifications
Granulocytes and Subsets, Monocytes, Myeloid Cells | |
1 x 109 Cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies EasySep™ Human CD33 Positive Selection Kit II