Learn More
STEMCELL Technologies EasySep™ Human CD138 Positive Selection Kit II
Immunomagnetic positive selection of human CD138+ cells (e.g. normal or malignant plasma cells for multiple myeloma research)
Brand: STEMCELL Technologies 17877
Description
- The EasySep™ Human CD138 Positive Selection Kit II is designed to isolate CD138+ (syndecan-1) cells from fresh or previously frozen bone marrow or peripheral blood mononuclear cells by positive selection
- Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD138 and dextran-coated magnetic particles
- This cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- The CD138 antigen is expressed on normal and malignant plasma cells (but not mature B cells)
- This product is an improved version of the EasySep™ Human CD138 Positive Selection Kit (Catalog #18357) that provides highly purified cells using a shorter protocol
Specifications
B Cells, Plasma | |
Cell Isolation | |
2 x 109 cells |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.