Learn More
STEMCELL Technologies EasySep™ HLA Whole Blood T Cell Enrichment Kit
Immunomagnetic isolation of untouched human T cells
Brand: STEMCELL Technologies 19951HLA
This item is not returnable.
View return policy
Description
- The Complete Processing Kit for whole blood uses EasySep™ HLA T Cell Enrichment Kit to isolate human T cells from HetaSep™-treated whole blood by negative selection
- Unwanted cells are targeted for removal with tetrameric antibody complexes recognizing non-T cells and dextran-coated magnetic particles
- The labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
T Cells | |
Cell Isolation | |
200 mL whole blood |