Learn More
STEMCELL Technologies EasySep™ HLA Whole Blood T Cell Enrichment Kit
Immunomagnetic isolation of untouched human T cells
Brand:  STEMCELL Technologies 19951HLA
Encompass
Product Code. 17177991
  • 10179.74 NOK / 1 set
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The Complete Processing Kit for whole blood uses EasySep™ HLA T Cell Enrichment Kit to isolate human T cells from HetaSep™-treated whole blood by negative selection
  • Unwanted cells are targeted for removal with tetrameric antibody complexes recognizing non-T cells and dextran-coated magnetic particles
  • The labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
Specifications
Specifications
T Cells
Cell Isolation
200 mL whole blood