Learn More
STEMCELL Technologies EasySep™ HLA B Cell Enrichment Kit
Column-free immunomagnetic isolation of untouched human B cells for HLA analysis
Brand:  STEMCELL Technologies 19054HLA
Encompass
Product Code. 17127901
  • 16227.06 NOK / 1 set
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ HLA B Cell Enrichment Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and dextran-coated magnetic particles
  • The labeled cells are separated using an EasySep™ magnet and without the use of columns
  • Desired cells are poured off into a new tube and are ready for serology or flow cytometry crossmatch assays
Specifications
Specifications
B Cells
Cell Isolation
1 x 109 Cells
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies EasySep™ HLA B Cell Enrichment Kit