Learn More
STEMCELL Technologies EasySep™ EasyStand™ x 6 with Magnets
Six linkable stands and six EasySep™ Magnets
Brand: STEMCELL Technologies 18137
This item is not returnable.
View return policy
Description
- The EasySep™ EasyStand™ facilitates the separation of one or more samples at one time
- Each EasySep™ EasyStand™ can hold a single EasySep™ Magnet (Catalog #18000), and a simple system links up to 6 individual EasySep™ EasyStands™ together
- After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes
Specifications
Cell Isolation | |
1 Ea. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.