Learn More
STEMCELL Technologies EasySep™ EasyStand™ x 6 with Magnets
Six linkable stands and six EasySep™ Magnets
Brand: STEMCELL Technologies 18137
Additional Details : Weight : 1.80000kg
Description
- The EasySep™ EasyStand™ facilitates the separation of one or more samples at one time
- Each EasySep™ EasyStand™ can hold a single EasySep™ Magnet (Catalog #18000), and a simple system links up to 6 individual EasySep™ EasyStands™ together
- After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes
Specifications
Cell Isolation | |
1 Ea. |