Learn More
STEMCELL Technologies EasySep™ Direct Human CD8+ T Cell Isolation Kit
Immunomagnetic isolation of CD8+ T cells directly from whole blood
Brand: STEMCELL Technologies 19663
This item is not returnable.
View return policy
Description
- The EasySep™ Direct Human CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from human whole blood by immunomagnetic negative selection
- The EasySep™ Direct procedure involves labeling RBCs, platelets and unwanted cells present in human whole blood with antibody complexes and magnetic particles
- The magnetically labeled RBCs, platelets and cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
Specifications
T Cells, T Cells, CD8+ | |
Cell Isolation | |
100 mL whole blood |