Learn More
STEMCELL Technologies EasySep™ Direct Human CD8+ T Cell Isolation Kit
Immunomagnetic isolation of CD8+ T cells directly from whole blood
Brand:  STEMCELL Technologies 19663
Encompass
Product Code. 17107951
  • 10666.89 NOK / 1 set
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Direct Human CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from human whole blood by immunomagnetic negative selection
  • The EasySep™ Direct procedure involves labeling RBCs, platelets and unwanted cells present in human whole blood with antibody complexes and magnetic particles
  • The magnetically labeled RBCs, platelets and cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
Specifications
Specifications
T Cells, T Cells, CD8+
Cell Isolation
100 mL whole blood