missing translation for 'onlineSavingsMsg'
Learn More

EAAT2 Mouse anti-Human, Unconjugated, Clone: 1C8D10, Proteintech

Mouse Monoclonal Antibody

1611.84 NOK - 4593.76 NOK

Specifications

Antigen EAAT2
Clone 1C8D10
Concentration 800 μg/mL
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16849505
View Documents Promotion Details
Proteintech
67083-1-IG-20UL
20 μL
1611.84 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16839505
View Documents Promotion Details
Proteintech
67083-1-IG-150UL
150 μL
4593.76 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The EAAT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EAAT2. This antibody recognizes human, mouse, rat antigen. The EAAT2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

EAAT2
800 μg/mL
Monoclonal
Mouse
6506
Primary
1C8D10
Western Blot, Immunohistochemistry
Unconjugated
Human, Rat, Mouse
IgG1
Videos
SDS
Documents

Documents

Product Certifications