missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ E2F6 (Human) Recombinant Protein
Human E2F6 full-length ORF ( AAH08348, 1 a.a. - 281 a.a.) recombinant protein with GST-tag at N-terminal.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH08348 |
---|---|
Gene ID (Entrez) | 1876 |
Name | E2F transcription factor 6 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16114301
|
Abnova™
H00001876-P01.10ug |
10 μg |
4115.00 NOK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16124301
|
Abnova™
H00001876-P01.25ug |
25 μg |
6245.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Sequence: MSQQRPARKLPSLLLDPTEETVRRRCRDPINVEGLLPSKIRINLEDNVQYVSMRKALKVKRPRFDVSLVYLTRKFMDLVRSAPGGILDLNKVATKLGVRKRRVYDITNVLDGIDLVEKKSKNHIRWIGSDLSNFGAVPQQKKLQEELSDLSAMEDALDELIKDCAQQLFELTDDKENERLAYVTYQDIHSIQAFHEQIVIAVKAPAETRLDVPAPREDSITVHIRSTNGPIDVYLCEVEQGQTSNKRSEGVGTSSSESTHPEGPEEEENPQQSEELLEVSN
Specifications
AAH08348 | |
E2F transcription factor 6 | |
125% SDS-PAGE Stained with Coomassie Blue | |
E2F-6, MGC111545 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
1876 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
E2F6 | |
GST |
Safety and Handling
missing translation for 'shelfLife' : Best use within three months from the date of receipt of this protein