Learn More
Thermo Scientific™ E1-ClipTip™ Electronic Pipette Charging Stand
Conveniently charge and store one or three pipettes with these custom charging stands.
Brand: Thermo Scientific™ 9420500
2190.10 NOK valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Description
Conveniently charge and store one or three E1-ClipTip pipettes with the Thermo Scientific™ E1-ClipTip™ Charging Stands.
Warranty: 1 year
Specifications
1 Pipette | |
Charging Stand | |
E1-ClipTip Electronic Single Channel Pipette | |
Universal power supply |
Promotions
-
Thermo Scientific ClipTip
,
Thermo Scientific ART
,
Thermo Scientific Finnpipette
,
Thermo Scientific Matrix
Exclusive Promotion on Liquid Handling
Unlock Precision: Top Pipettes and Tips on Promotion!Promo Code: 25076Expires: 2025-06-30
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
-
Invitrogen
,
ABI Applied Biosystems
,
Thermo Scientific
,
Affymetrix eBioscience
Nurture Your Progress
Exclusive savings to help your research thrive.Promo Code: 24108Expires: 2025-07-04