Learn More
3M™ E-A-R™ UltraTech Earplugs
3M™ E-A-R™ UltraTech Earplugs greatly improve the ability to perceive speech, warning signals and machinery noise whilst reducing harmful noise levels effectively.
Brand:  3M™ 7000103799
Product Code. 17217630
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Improve wearer's ability to perceive speech, warning signals and machinery noise whilst reducing harmful noise levels effectively
  • Earplug are ideal for musicians who require a flat attenuation protection
  • Conical shape plugs help provide effective seal for reassured protection
  • Effective protection against moderate levels of harmful noise SNR 21 dB
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black, White
21 dB
No
1 Pair/Box
Uncorded
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
3M™ E-A-R™ UltraTech Earplugs > Color: Black, White