Learn More
3M™ E-A-R™ Ultrafit™ Ear Plugs Product Code.: 15799847
Add to basket
3M™ E-A-R™ Ultrafit™ Ear Plugs
Product Code. 15799847
Color: Yellow, Blue
Click to view available options
Color: Yellow, Blue
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 15799847
Brand: 3M™ 7000103727
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
UltraFit™ reusable ear plugs are durable and feature a triple-flange, push-to-fit design to provide comfort and convenience. Easy to wash with soap and water so they can be used multiple times, reducing waste.
  • Push-to-fit style for easy and hygienic fit
  • Universal size fits comfortably in most ear canals
  • Premolded, triple-flange ear plug design inserts easily
  • No roll-down necessary
  • Soft polymer is washable and reusable
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Size Universal
Packaging Type Carrying case
Pairs per Package 50 Pairs/Pk.
Plug Material Elastomeric polymer
Noise Reduction Rating (NRR) 32 dB
Corded Corded
Plug Shape Flanged
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
3M™ E-A-R™ Ultrafit™ Ear Plugs >