Description
- Set the standard in hearing protection as the world's first foam earplugs
- Firm material with slow recovery time helps achieve good fit
- Moisture-resistant material helps maintain a consistent expansion time, even in humid environments
- Low equilibrium pressure helps reduce pressure in ear canal thus increasing comfort and wearability
- Exposed cell surface texture resists movement and helps maintain effective seal
- Excellent sound attenuation characteristics
- Supplied in re-sealable pillow-pack for ease of use
- Sweat and humidity resistant which helps improve comfort- ideally suited for use in hot and humid conditions
Specifications
Specifications
Color | Yellow |
Certifications/Compliance | CE Approved |
Packaging Type | Polybag |
Pairs per Package | 200 Pairs/Bag |
Noise Reduction Rating (NRR) | 28 dB |
Corded | Uncorded |
Product Type | Ear Plugs |
Dielectric | No |
Material | PVC |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ E-A-R™ Classic™ Earplugs PB-01-000 >