Learn More
3M™ E-A-R™ Classic™ Earplugs
Provides excellent hearing protection and all day comfort. 3M E-A-R Classic Earplug offer either 28 or 29dB.
915.00 NOK - 1500.00 NOK
Specifications
Color | Yellow |
---|---|
Certifications/Compliance | CE Approved |
Noise Reduction Rating (NRR) | 28db |
Product Type | Classic Earplug |
Style | Roll-down |
Product Code | Brand | Corded | Noise Reduction Rating (NRR) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Corded | Noise Reduction Rating (NRR) | Price | Quantity & Availability | |||||
12354749
|
3M™
7000052844 |
Corded | 28db |
1500.00 NOK
200 pairs |
Estimated Shipment: 25-06-2025
Log in to see stock. |
|||||
12344749
|
3M™
7000038198 |
Uncorded | 28db |
915.00 NOK
250 pairs |
Estimated Shipment: 25-06-2025
Log in to see stock. |
|||||
Description
Made from a soft, energy-absorbing polymer foam that provides excellent hearing protection and all day comfort. Ideal for use where noise is a problem, either in work or leisure activities.- Firm material with slow recovery time helps achieve good fit
- Special cylindrical shape that fits most ear canals and provides reliable seal
- Low equilibrium pressure helps reduce pressure in ear canal thus increasing comfort and wearability
- One size fits majority of wearers
- Excellent sound attenuation characteristics
- Sweat and humidity resistant which helps improve comfort
- Ideally suited for use in hot and humid conditions
Specifications
Yellow | |
28db | |
Roll-down |
CE Approved | |
Classic Earplug |
Product Title
3M™ E-A-R™ Classic™ Earplugs