missing translation for 'onlineSavingsMsg'
Learn More

Dynamitin (1-83 AA) Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen Dynamitin (1-83 AA)
Concentration 0.33 mg/mL
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16846183
missing translation for 'viewDocuments'
Proteintech
10031-2-AP-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16836183
missing translation for 'viewDocuments'
Proteintech
10031-2-AP-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is against the N terminal 59aa of full length.

This gene encodes a 50-kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kDa. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 4-5 copies per dynactin molecule. It contains three short alpha-helical coiled-coil domains that may mediate association with self or other dynactin subunits. It may interact directly with the largest subunit (p150) of dynactin and may affix p150 in place.
Specifications

Specifications

Dynamitin (1-83 AA)
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Mouse, Human
Q13561, Q99KJ8
10540, 69654
IgG
Antigen Affinity Chromatography
Antibody
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
DCTN 50, DCTN2, DCTN50, dynactin 2, dynactin 2 (p50), Dynactin subunit 2, DYNAMITIN, p50 dynamitin, RBP50
Dctn2
Primary
-20°C
DCTN2
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.