Learn More
Abeba™ Dynamic 37313 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand:  Abeba™ 37313/43
 View more versions of this product
Product Code. 15680868
  • 733.00 NOK / 1 pair
Only null left
Estimated Shipment: 31-07-2025
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather white/red
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
37313
White
43
Polyurethane
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Abeba™ Dynamic 37313 Shoes > Size: 43