Learn More
Invitrogen™ Dynabeads™ RNA Binding Buffer
Dynabeads RNA Purification Buffer is intended for use with Dynabeads Carboxylic Acid for RNA Purification, providing a simple, scalable, and automation-ready solution for purification of crude RNA produced by in vitro transcription (IVT).
606.00 NOK - 1614.00 NOK
Specifications
Content And Storage | Store at 2°C to 8°C and protected from light. |
---|---|
Isolation Technology | Magnetic Bead |
Purification Time | Variable |
Sample Type | Crude RNA |
For Use With (Equipment) | KingFisher™ Sample Purification System, DynaMag™ magnets |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
17847378
|
Invitrogen™
65040D |
20 mL |
606.00 NOK
20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17804058
|
Invitrogen™
65042D |
50 mL |
1614.00 NOK
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dynabeads RNA Purification Buffer is intended for use with Dynabeads Carboxylic Acid for RNA Purification, providing a simple, scalable, and automation-ready solution for purification of crude RNA produced by in vitro transcription (IVT). The combination is particularly suitable where flexibility and high purity are required, such as in discovery phase of vaccine development and following the solid-phase IVT reaction with Dynabeads Streptavidin for In Vitro Transcription.
This buffer combined with Dynabeads Carboxylic Acid for RNA Purification offer efficiency, scalability (micrograms to grams of mRNA) in manual and automated workflows, and quality to navigate from RNA-based drug discovery to commercial manufacturing without incurring extra time or costs from switching technology or raw material.
Crude RNA from an IVT reaction is mixed with the beads and the buffer is added. The RNA binds to the bead surface and the remaining components of the reaction mixture are removed by applying a magnet and discarding the supernatant. Scripts are available for automated purification workflows with KingFisher systems.
The buffer is offered here as a stand-alone product. It is also included in the kits below:
Dynabeads RNA Purification Kit (Cat. No. 65030D)
Dynabeads In Vitro Transcription and RNA Purification Kit (Cat. No. 65034D)
To learn more about other products and support documentation available for GMP-compliant manufacturing of mRNA vaccines and therapeutics visit: thermofisher.com/dynabeadsmrna
Specifications
Store at 2°C to 8°C and protected from light. | |
Variable | |
KingFisher™ Sample Purification System, DynaMag™ magnets | |
Dynabeads™ | |
Ambient Temperature |
Magnetic Bead | |
Crude RNA | |
In Vitro Transcription, Synthetic RNA, Crude RNA Purification (from IVT Reaction Mix) | |
No detectable activity level | |
RNA |
Safety and Handling
missing translation for 'shelfLife' : 18 Months (from date of manufacture)
Your input is important to us. Please complete this form to provide feedback related to the content on this product.