Learn More
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels with Thermostatic Jacket
Borosilicate glass reaction vessel with thermostatic jacket
Brand: Lenz Laborglasinstrumente™ 06010276
8489.98 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
DURAN™ tubing, cylindrical shape with graduation and laboratory flange LF. Thermostatic jacket with either GL 14, GL 18, KF 15 or KF 25 connectors.Specifications
3 L | |
325 mm | |
Reaction vessel | |
Flat |
Borosilicate Glass, Duran Glass | |
Thermostatic jacket | |
Cylindrical |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels with Thermostatic Jacket > Capacity: 3L; Height: 325mm