Learn More
Lenz Laborglasinstrumente™ Duran™ Nitrogen Tube
Brand:  Lenz Laborglasinstrumente™ 03370222
 View more versions of this product
Product Code. 10737141
  • 819.00 NOK / Each
Only null left
Estimated Shipment: 04-08-2025
Specifications
25 mL
Nitrogen Tube
1 Pack
Duran Glass
Cone and NS stopcock
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Duran™ Nitrogen Tube > Capacity: 25mL