Learn More
Lenz Laborglasinstrumente™ Duran™ Nitrogen Tube
Brand:  Lenz Laborglasinstrumente™ 03370237
 View more versions of this product
Product Code. 10366211
  • 867.00 NOK / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Specifications
100 mL
Nitrogen Tube
1 Pack
Duran Glass
Cone and NS stopcock
Product Title
Lenz Laborglasinstrumente™ Duran™ Nitrogen Tube > Capacity: 100mL