Learn More
Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks
Manufactured from Duran™ glass. Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks include a conical ground joint.
145.00 NOK - 2400.00 NOK
Specifications
Certifications/Compliance | DIN EN ISO 4797 |
---|---|
Material | Borosilicate Glass, Duran Glass |
Neck Style | Medium |
Bottom Shape | Round |
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Standard Taper Joint | Price | Quantity & Availability | |||||
10483861
|
Lenz Laborglasinstrumente™
03002970 |
1000 mL | 131 mm | NS 29/32 |
2400.00 NOK
Pack of 10 |
Estimated Shipment: 12-08-2025
Log in to see stock. |
|||||
15360895
|
Lenz Laborglasinstrumente™
03001413 |
10 mL | 31 mm | NS 14/23 |
996.00 NOK
Pack of 10 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||||
10559421
|
Lenz Laborglasinstrumente™
03002937 |
100 mL | 64 mm | NS 29/32 | N/A | ||||||
11737425
|
Lenz Laborglasinstrumente™
03002937 |
100 mL | 64 mm | NS 29/32 |
943.00 NOK
Pack of 10 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||||
10589611
|
Lenz Laborglasinstrumente™
03002973 |
2000 mL | 166 mm | NS 29/32 | N/A | ||||||
10211551
|
Lenz Laborglasinstrumente™
03002949 |
250 mL | 85 mm | NS 29/32 |
1800.00 NOK
Pack of 10 |
Estimated Shipment: 04-08-2025
Log in to see stock. |
|||||
15370885
|
Lenz Laborglasinstrumente™
03001422 |
25 mL | 45 mm | NS 14/23 |
537.00 NOK
Pack of 10 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
10220831
|
Lenz Laborglasinstrumente™
03002958 |
500 mL | 105 mm | NS 29/32 | N/A | ||||||
11915956
|
Lenz Laborglasinstrumente™
03002958 |
500 mL | 105 mm | NS 29/32 |
2180.00 NOK
Pack of 10 |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
10710992
|
Lenz Laborglasinstrumente™
03001428 |
50 mL | 51 mm | NS 14/23 |
685.00 NOK
Pack of 10 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
|||||
10508851
|
Lenz Laborglasinstrumente™
03002928 |
50 mL | 51 mm | NS 29/32 | N/A | ||||||
Description
- Manufactured from Duran™ glass
- Complies with DIN EN ISO 4797 standards
- Includes conical ground joint
Specifications
DIN EN ISO 4797 | |
Borosilicate Glass, Duran Glass | |
Medium | |
Round |
Product Title
Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks