Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Coil Condenser
Duran™ Glass Coil Condensers made of DURAN™ tubing.
Brand: Lenz Laborglasinstrumente™ 05250204
This item is not returnable.
View return policy
Description
- Duran™ Glass
Specifications
DIN 12593 and in addition to DIN | |
For condensing vapors | |
1 Ea. | |
Duran Glass |
Cooling Surface: 240 cm2 | |
160 mm | |
NS 29/32 | |
Coil Condenser |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Duran™ Glass Coil Condenser > Neck Size: 29/32, Jacket Length: 160 mm