Learn More
Lenz Laborglasinstrumente™ Duran™ Gas Sampling Tube
Brand:  Lenz Laborglasinstrumente™ 05680058
 View more versions of this product
Product Code. 10057970
  • 1245.00 NOK / Each
Only null left
Estimated Shipment: 25-07-2025
Specifications
500 mL
Gas Sampling Tube
DIN 12473-1
Duran Glass
Nozzle GL14 Thread and septa
1 Pack
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Duran™ Gas Sampling Tube > Capacity: 500mL