Learn More
Lenz Laborglasinstrumente™ Duran™ Extractors
Brand:  Lenz Laborglasinstrumente™ 05360037
 View more versions of this product
Product Code. 10615051
  • 2970.00 NOK / Each
Only null left
Estimated Shipment: 19-08-2025
Specifications
Dimroth Condenser
100 mL
Dimroth condenser
GL 14
Extractor
Duran Glass
For the extraction from solids with solvents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Duran™ Extractors > Capacity: 100mL