Learn More
Lenz Laborglasinstrumente™ Erlenmeyer Flasks With Conical Ground Joint
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
21.63 NOK - 3540.00 NOK
Specifications
Material | Borosilicate Glass, Duran Glass |
---|---|
Includes | With scale (approx. volume/mL), with inscription label |
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||||
15340895
|
Lenz Laborglasinstrumente™
03022970 |
1000 mL | NS29/32 |
3540.00 NOK
Pack of 10 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
10298070
|
Lenz Laborglasinstrumente™
03021937 |
100 mL | NS19/26 | N/A | N/A | N/A | |||||
12034098
|
Lenz Laborglasinstrumente™
03021937 |
100 mL | NS19/26 |
1210.00 NOK
Pack of 10 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
10662351
|
Lenz Laborglasinstrumente™
03022937 |
100 mL | NS29/32 |
1050.00 NOK
Pack of 10 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
15390885
|
Lenz Laborglasinstrumente™
03022946 |
200 mL | NS29/32 |
1875.00 NOK
Pack of 10 |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
10741434
|
Lenz Laborglasinstrumente™
03022949 |
250 mL | NS29/32 |
1085.00 NOK
Pack of 10 |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
10017750
|
Lenz Laborglasinstrumente™
03022952 |
300 mL | NS29/32 | N/A | N/A | N/A | |||||
11965976
|
Lenz Laborglasinstrumente™
03022952 |
300 mL | NS29/32 |
1200.00 NOK
Pack of 10 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
10434061
|
Lenz Laborglasinstrumente™
03022958 |
500 mL | NS29/32 |
1175.00 NOK
Pack of 10 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
10533831
|
Lenz Laborglasinstrumente™
03021928 |
50 mL | NS19/26 |
993.00 NOK
Pack of 10 |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
10652351
|
Lenz Laborglasinstrumente™
03022928 |
50 mL | NS29/32 | N/A | N/A | N/A | |||||
12163991
|
Lenz Laborglasinstrumente™
03022928 |
50 mL | NS29/32 |
925.00 NOK
Pack of 10 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
Description
- Acc. to DIN EN ISO 4797
- With scale (approx. volume/mL)
- With inscription label
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Borosilicate Glass, Duran Glass | |
With scale (approx. volume/mL), with inscription label |
Product Title
Lenz Laborglasinstrumente™ Erlenmeyer Flasks With Conical Ground Joint