Learn More
DWK Life Sciences DURAN™ Bloom Test Vessel
The Bloom test glass is used to determine the gelatinous strength (determination of the Bloom value).
Brand:  DWK Life Sciences 211250105
6867.41 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 15241649
  • 8945.00 NOK / Pack of 10
In Stock
to see stock.
Explore more special offers
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. In the fields of food technology and pharmaceutical technology, the gelling power of gelatine (gelatine strength) is described. For this purpose, the gelatine is poured into a specific shape, which is used to determine the Bloom value. A Bloom test glass is required for this step.
  • Thick-walled and therefore particularly resistant to mechanical stress
TRUSTED_SUSTAINABILITY
Specifications
Specifications
59 mm
Performing a Bloom test to measure the strength of a gel or gelatin
Autoclavable
Borosilicate Glass (3.3)
10/Pk.
Product Title
DWK Life Sciences DURAN™ Bloom Test Vessel > 59 mm