Learn More
Lenz Laborglasinstrumente™ Duran Adapters with NS Stopcock
Manufactured from DURAN™ glass tubing. Lenz Laborglasinstrumente™ Duran™ Adapters with NS Stopcock includes bent glass olive (90°) and NS stopcock with solid glass plug, bore: 2.5mm.
Brand:  Lenz Laborglasinstrumente™ 05098004
 View more versions of this product
Product Code. 11816144
  • 456.00 NOK / Each
Only null left
Estimated Shipment: 17-07-2025
Description
  • Manufactured from DURAN™ glass tubing
  • Includes bent glass olive (90°) and stopcock with solid glass plug, bore: 2.5mm.
Specifications
Specifications
NS 29/32
Bent glass olive (90°), Solid glass plug, bore 2.5mm
Duran Glass
Adapter
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Duran Adapters with NS Stopcock > Bore Size: 2.5mm; NS Cone Size: 29/32