Learn More
Vileda Professional™ DuoPress™ Wringer
The CE Trolley range has been specifically designed for use in controlled environments where maximum contamination control is needed. This DuoPress™ wringer is part of the CE Bucket Trolley line for wringing of flat mops.
Brand: Vileda Professional™ 168207
Description
- For use on the CE Bucket Trolley with the bucket method
- 100% polypropylene chassis and stainless steel lever
- Effectively wrings CE Duo™ mops
- Ergonomic lever for maximum pressure with minimal effort
- Base of press support dropped 10 mm to create additional space for mops, making it easier to wring
Specifications
Mop Wringer | |
Polypropylene and Stainless steel | |
Wringer base, Support plate and Wringer | |
1 Ea. |