missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTNB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12045-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes dystrobrevin beta, a component of the dystrophin-associated protein complex (DPC). The DPC consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and dystrobrevin alpha and beta. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Dystrobrevin beta is thought to interact with syntrophin and the DP71 short form of dystrophin. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
DTNB | |
Polyclonal | |
Unconjugated | |
DTNB | |
Beta dystrobrevin, DTN B, DTNB, Dystrobrevin beta, dystrobrevin, beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13528, 1838, 362715 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60941, O70585, P84060 | |
DTNB | |
DTNB Fusion Protein Ag2675 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |