Learn More
KGW Isotherm™ DSS Stainless Steel Dewar Flasks
4190.00 NOK - 7395.00 NOK
Specifications
Material | DSS Stainless Steel |
---|---|
For Use With (Application) | Used to Store and Transport liquid nitrogen ( LN2 ), Dry Ice ( CO2 ) and Other Coolants |
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | |||||
10722023
|
KGW Isotherm™
2101 |
1 L | 225 mm |
4190.00 NOK
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10065171
|
KGW Isotherm™
2102 |
1 L | 178 mm |
4475.00 NOK
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||||
10594502
|
KGW Isotherm™
2103 |
2 L | 305 mm |
5115.00 NOK
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
10228511
|
KGW Isotherm™
2104 |
3 L | 190 mm |
7395.00 NOK
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
Description
Lid is sold separately; see KGW Isotherm Lids (10166941, 10543932, 10545082).
Specifications
DSS Stainless Steel | |
Used to Store and Transport liquid nitrogen ( LN2 ), Dry Ice ( CO2 ) and Other Coolants |
Product Title
KGW Isotherm™ DSS Stainless Steel Dewar Flasks