missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsRNA Antibody (J5), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11396C
This item is not returnable.
View return policy
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, ImmunoblotSpecifications
dsRNA | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Broad Species Reactivity | |
Purified |
ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot | |
J5 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting | |
Double-stranded RNA | |
Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone. | |
0.2 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction